2,4-Dichloro-5-methylaniline 98%

Supplier: Ambeed
Warning

A882660-10G A882660-25G A882660-5G
77256-472EA 50.47 USD
77256-472 77256-474 77256-476
2,4-Dichloro-5-methylaniline 98%
2,4-Dichloro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
MDL Number: MFCD09833435
CAS Number: 17601-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR