ω,ω-Difluoroacetophenone 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2-Difluoroacetophenone, 2,2-Difluoro-1-phenylethanone, α,α-Difluoroacetophenone
A880757-100MG
A880757-250MG
A880757-5G
A880757-1G
77256-466EA
19.57
USD
77256-466
77256-468
77256-470
77569-610
ω,ω-Difluoroacetophenone 95%
ω,ω-Difluoroacetophenone
Precaución: For research use only.
Formula:
C₈H₆F₂O MW: 156.13 g/mol |
MDL Number:
MFCD06246879 CAS Number: 395-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...