Methyl 2-(3-amino-4-bromophenoxy)acetate 97%

Supplier: Ambeed
Warning

A880075-100MG A880075-1G A880075-250MG
77244-636EA 48 USD
77244-636 77244-638 77244-640
Methyl 2-(3-amino-4-bromophenoxy)acetate 97%
Methyl 2-(3-amino-4-bromophenoxy)acetate

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₃
MDL Number: MFCD22378163
CAS Number: 1387563-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR