2,2'-Dihydroxybenzophenone 99%

Supplier: Ambeed
Warning

Synonyms: Bis(2-hydroxyphenyl)methanone

A878182-100G A878182-1G A878182-25G A878182-5G
77256-458EA 953.78 USD
77256-458 77256-460 77256-462 77256-464
2,2'-Dihydroxybenzophenone 99%
2,2'-Dihydroxybenzophenone

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₃
MW: 214.2167 g/mol
MDL Number: MFCD00002217
CAS Number: 835-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR