Disodium diphenylsulfone-4,4'-dichloro-3,3'-disulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Sodium 5,5'-sulphonylbis(2-chlorobenzenesulphonate), Diphenylsulphone-4,4'-dichloro-3,3'-disulphonic acid disodium salt

A877845-1G A877845-250MG A877845-5G
77244-622EA 23.69 USD
77244-622 77244-624 77244-626
Disodium diphenylsulfone-4,4'-dichloro-3,3'-disulfonate 98%
Disodium diphenylsulfone-4,4'-dichloro-3,3'-disulfonate

Precaución: for Research Use Only

Formula: C₁₂H₈Cl₂O₈S₃
MW: 491.23 g/mol
MDL Number: MFCD13194854
CAS Number: 51698-33-0
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR