2-(Dimethylaminomethyl)-3-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((Dimethylamino)methyl)pyridin-3-ol
A877700-100G
A877700-10G
A877700-1G
A877700-25G
A877700-5G
77256-448EA
311
USD
77256-448
77256-450
77256-452
77256-454
77256-456
2-(Dimethylaminomethyl)-3-hydroxypyridine 98%
2-(Dimethylaminomethyl)-3-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₈H₁₂N₂O MW: 152.2 g/mol |
MDL Number:
MFCD00006347 CAS Number: 2168-13-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...