4-Chloro-3-fluorotoluene 98%

Supplier: Ambeed
Danger

A875388-10G A875388-25G A875388-5G A875388-1G
77294-100EA 16 USD
77294-100 77294-102 77294-104 77308-478
4-Chloro-3-fluorotoluene 98%
4-Chloro-3-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆ClF
MDL Number: MFCD00143295
CAS Number: 5527-94-6
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR