2,2'-Methylenedianiline 95%

Supplier: Ambeed
Warning

A875278-100MG A875278-1G A875278-250MG
77244-604EA 17 USD
77244-604 77244-606 77573-768
2,2'-Methylenedianiline 95%
2,2'-Methylenedianiline

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂
MDL Number: MFCD01109641
CAS Number: 6582-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR