(2R,3R,4S,5S,6R)-2-Fluoro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 97%

Supplier: Ambeed
Warning

A874998-100MG A874998-1G A874998-250MG A874998-5G
77244-596EA 7 USD
77244-596 77244-598 77244-600 77244-602
(2R,3R,4S,5S,6R)-2-Fluoro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 97%
(2R,3R,4S,5S,6R)-2-Fluoro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₆H₁₁FO₅
MDL Number: MFCD00132903
CAS Number: 2106-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR