3',4'-Dimethylacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Dimethylphenyl)ethan-1-one, 1-(3,4-Dimethylphenyl)ethanone

A874898-1G A874898-25G
77281-824EA 9.27 USD
77281-824 77281-826
3',4'-Dimethylacetophenone 98%
3',4'-Dimethylacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00008743
CAS Number: 3637-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR