2,7-Dimethylpyrene 95%

Supplier: Ambeed
Warning

A874887-100MG A874887-1G A874887-250MG
77256-442EA 176 USD
77256-442 77256-444 77256-446
2,7-Dimethylpyrene 95%
2,7-Dimethylpyrene

Precaución: for Research Use Only

Formula: C₁₈H₁₄
MDL Number: MFCD02683750
CAS Number: 15679-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR