2,4,6-Triphenylpyrilium tetrafluoroborate 95%

Supplier: Ambeed
Danger

A873813-100G A873813-10G A873813-1G A873813-25G A873813-5G
77256-418EA 275 USD
77256-418 77256-420 77256-422 77256-424 77256-426
2,4,6-Triphenylpyrilium tetrafluoroborate 95%
2,4,6-Triphenylpyrilium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₂₃H₁₇BF₄O
MDL Number: MFCD00012001
CAS Number: 448-61-3
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR