Isophorone oxide 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,6-Trimethyl-7-oxabicyclo[4.1.0]heptan-2-one, 2,3-Epoxy-3,5,5-trimethyl-1-cyclohexanone, 3,5,5-Trimethyl-2,3-epoxy-1-cyclohexanone

A872972-25G A872972-5G
77294-096EA 28.84 USD
77294-096 77294-098
Isophorone oxide 98%
Isophorone oxide

Precaución: for Research Use Only

Formula: C₉H₁₄O₂
MW: 154.21 g/mol
MDL Number: MFCD00085465
CAS Number: 10276-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR