3-(2,4-Dimethylphenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4-Dimethylphenyl)propanoic acid

A872461-100MG A872461-250MG A872461-5G A872461-1G
77281-810EA 10.3 USD
77281-810 77281-812 77281-814 77749-677
3-(2,4-Dimethylphenyl)propionic acid 97%
3-(2,4-Dimethylphenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00796498
CAS Number: 1811-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR