(R)-1-{(R)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine 98%

Supplier: Ambeed
Warning

A872410-100MG A872410-1G A872410-250MG A872410-5G
77244-586EA 22 USD
77244-586 77244-588 77244-590 77244-592
(R)-1-{(R)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine 98%
(R)-1-{(R)-2-[Bis[3,5-bis(trifluoromethyl)phenyl]phosphino]ferrocenyl}ethyldi(3,5-xylyl)phosphine

Precaución: for Research Use Only

Formula: C₄₄H₃₆F₁₂FeP₂
MDL Number: MFCD08561153
CAS Number: 166172-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR