Methyl 4-(morpholin-4-yl)benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-morpholinobenzoate

A871959-1G A871959-250MG A871959-5G
77294-090EA 57.68 USD
77294-090 77294-092 77294-094
Methyl 4-(morpholin-4-yl)benzoate 98%
Methyl 4-(morpholin-4-yl)benzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.252 g/mol
MDL Number: MFCD03012127
CAS Number: 23676-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR