(4,6-Dimethoxypyridin-3-yl)boronic acid 97%

Supplier: Ambeed
Warning

A871956-100MG A871956-1G A871956-250MG
77256-412EA 84 USD
77256-412 77256-414 77256-416
(4,6-Dimethoxypyridin-3-yl)boronic acid 97%
(4,6-Dimethoxypyridin-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄
MDL Number: MFCD12964580
CAS Number: 915021-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR