3-(Cyclopentyloxy)-4-methoxybenzaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Cyclopentyloxy)-4-formylanisole

A871954-1G A871954-25G A871954-5G
77281-804EA 19.57 USD
77281-804 77281-806 77281-808
3-(Cyclopentyloxy)-4-methoxybenzaldehyde 98%
3-(Cyclopentyloxy)-4-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₆O₃
MW: 220.264 g/mol
MDL Number: MFCD00673136
CAS Number: 67387-76-2
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR