(S)-2-Amino-3-(4-((5-amino-2-phenylbenzo[d]oxazol-7-yl)methoxy)-3,5-dichlorophenyl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: JPH203

A871573-100MG A871573-10MG A871573-1MG A871573-25MG A871573-2MG A871573-50MG
77268-694EA 347.11 USD
77268-694 77268-696 77268-698 77268-700 77268-702 77618-446
(S)-2-Amino-3-(4-((5-amino-2-phenylbenzo[d]oxazol-7-yl)methoxy)-3,5-dichlorophenyl)propanoic acid 98%
(S)-2-Amino-3-(4-((5-amino-2-phenylbenzo[d]oxazol-7-yl)methoxy)-3,5-dichlorophenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₃H₁₉Cl₂N₃O₄
MW: 472.32 g/mol
MDL Number: MFCD30534398
CAS Number: 1037592-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR