(R)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate 98%

Supplier: Ambeed
Warning

A871130-100MG A871130-250MG
77256-406EA 70 USD
77256-406 77256-408
(R)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate 98%
(R)-3,3'-Bis[4-(1-naphthalenyl)phenyl]-1,1'-binaphthyl-2,2'-diyl Hydrogenphosphate

Precaución: for Research Use Only

Formula: C₅₂H₃₃O₄P
CAS Number: 1450925-06-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR