3-Bromo-5-fluoro-4-methoxyaniline 98%

Supplier: Ambeed
Danger

A870537-1G A870537-250MG A870537-5G
77281-798EA 49.44 USD
77281-798 77281-800 77281-802
3-Bromo-5-fluoro-4-methoxyaniline 98%
3-Bromo-5-fluoro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFNO
MW: 220.04 g/mol
MDL Number: MFCD08062384
CAS Number: 875664-44-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR