3-Methoxyphenylacetone 97%

Supplier: Ambeed
Warning

Synonyms: 1-(3-Methoxyphenyl)propan-2-one

A870340-10G A870340-1G A870340-250MG A870340-25G A870340-5G
77281-788EA 174.07 USD
77281-788 77281-790 77281-792 77281-794 77281-796
3-Methoxyphenylacetone 97%
3-Methoxyphenylacetone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00008771
CAS Number: 3027-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR