4-Methyl-2-(methylthio)pyrimidine 97%

Supplier: Ambeed
Danger

Synonyms: 4-Methyl-2(methylsulphanyl)pyrimidine

A870173-10G A870173-1G A870173-25G A870173-5G
77294-082EA 57.68 USD
77294-082 77294-084 77294-086 77294-088
4-Methyl-2-(methylthio)pyrimidine 97%
4-Methyl-2-(methylthio)pyrimidine

Precaución: for Research Use Only

Formula: C₆H₈N₂S
MW: 140.21 g/mol
MDL Number: MFCD00023242
CAS Number: 14001-63-9
UN: 3334
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR