trans-4-Nitrocinnamaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: trans-3-(4-Nitrophenyl)-2-propenal, (E)-3-(4-Nitrophenyl)acrylaldehyde, (E)-4-Nitrocinnamaldehyde, (E)-3-(4-Nitrophenyl)-2-propenal

A869879-1G A869879-250MG A869879-5G
77294-076EA 49.44 USD
77294-076 77294-078 77294-080
trans-4-Nitrocinnamaldehyde 97%
trans-4-Nitrocinnamaldehyde

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD00007379
CAS Number: 49678-08-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR