3,5-Dibromo-1-(trimethylsilyl)benzene 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dibromophenyltrimethylsilane, (3,5-Dibromophenyl)trimethylsilane

A868665-1G A868665-250MG
77281-776EA 41.2 USD
77281-776 77281-778
3,5-Dibromo-1-(trimethylsilyl)benzene 97%
3,5-Dibromo-1-(trimethylsilyl)benzene

Precaución: for Research Use Only

Formula: C₉H₁₂Br₂Si
MW: 308.0851 g/mol
MDL Number: MFCD03844787
CAS Number: 17878-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR