3-(2-Cyanoethylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-((2-Cyanoethyl)carbamoyl)phenyl)boronic acid, 3-(2-Cyanoethylaminocarbonyl)benzeneboronic acid

A868444-1G A868444-250MG A868444-5G
77281-770EA 82.4 USD
77281-770 77281-772 77281-774
3-(2-Cyanoethylcarbamoyl)phenylboronic acid 98%
3-(2-Cyanoethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BN₂O₃
MW: 218.0200 g/mol
MDL Number: MFCD04115697
CAS Number: 762262-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR