(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-DMM-Garphos, (R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxybiphenyl, (R)-(4,4',6,6'-Tetramethoxybiphenyl-2,2'-diyl)bis[bis(4-methoxy-3,5-dimethylphenyl)phosphine]

A868409-100MG A868409-250MG
77256-402EA 82.4 USD
77256-402 77256-404
(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl ≥98%, ee 99%
(R)-2,2'-Bis[bis(4-methoxy-3,5-dimethylphenyl)phosphino]-4,4',6,6'-tetramethoxy)-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₅₂H₆₀O₄P₂
MW: 810.970 g/mol
MDL Number: MFCD19443626
CAS Number: 1365531-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR