2-Bromo-6-hydrazinylpyridine 98%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-6-hydrazinopyridine

A868007-100G A868007-10G A868007-1G A868007-25G
77268-666EA 183.34 USD
77268-666 77268-668 77268-670 77268-672
2-Bromo-6-hydrazinylpyridine 98%
2-Bromo-6-hydrazinylpyridine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.028 g/mol
MDL Number: MFCD09743754
CAS Number: 26944-71-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR