2-(((1R,2R,3aS,9aS)-2-Hydroxy-1-((S)-3-hydroxyoctyl)-2,3,3a,4,9,9a-hexahydro-1H-cyclopenta[b]naphthalen-5-yl)oxy)acetic acid, sodium salt 97%

Supplier: Ambeed
Danger

A867461-10MG A867461-1MG A867461-25MG A867461-5MG A867461-100MG A867461-50MG A867461-250MG
77244-560EA 129 USD
77244-560 77244-562 77244-564 77244-566 77618-444 77747-636 77747-635
2-(((1R,2R,3aS,9aS)-2-Hydroxy-1-((S)-3-hydroxyoctyl)-2,3,3a,4,9,9a-hexahydro-1H-cyclopenta[b]naphthalen-5-yl)oxy)acetic acid, sodium salt 97%
2-(((1R,2R,3aS,9aS)-2-Hydroxy-1-((S)-3-hydroxyoctyl)-2,3,3a,4,9,9a-hexahydro-1H-cyclopenta[b]naphthalen-5-yl)oxy)acetic acid, sodium salt

Precaución: for Research Use Only

Formula: C₂₃H₃₃NaO₅
MDL Number: MFCD08141832
CAS Number: 289480-64-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR