4-Bromo-2,5-difluoroanisole 99%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2,5-difluoro-4-methoxybenzene

A866944-1G A866944-5G
77294-072EA 14.42 USD
77294-072 77294-074
4-Bromo-2,5-difluoroanisole 99%
4-Bromo-2,5-difluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00143258
CAS Number: 202865-60-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR