4-Bromo-2,3,5,6-tetrafluorobenzonitrile 98%

Supplier: Ambeed
Warning

A865158-10G A865158-1G A865158-250MG A865158-25G A865158-5G
77294-060EA 260.59 USD
77294-060 77294-062 77294-064 77294-066 77294-068
4-Bromo-2,3,5,6-tetrafluorobenzonitrile 98%
4-Bromo-2,3,5,6-tetrafluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇BrF₄N
MW: 253.9792 g/mol
MDL Number: MFCD00075282
CAS Number: 17823-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR