2,3-Dihydro-2-methylbenzofuran 98%

Supplier: Ambeed
Warning

A865020-1G A865020-250MG A865020-5G
77256-394EA 39 USD
77256-394 77256-396 77256-398
2,3-Dihydro-2-methylbenzofuran 98%
2,3-Dihydro-2-methylbenzofuran

Precaución: for Research Use Only

Formula: C₉H₁₀O
MDL Number: MFCD00023003
CAS Number: 1746-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR