(4S,4'S)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%

Supplier: Ambeed
Warning

A864770-100MG A864770-250MG
77256-392EA 54.59 USD
77256-392 77747-633
(4S,4'S)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole) 98%
(4S,4'S)-2,2'-(Pentane-3,3-diyl)bis(4-phenyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₂
MW: 362.46 g/mol
MDL Number: MFCD30187345
CAS Number: 190791-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR