TAK-659 Hydrochloride 98+%

Supplier: Ambeed
Warning

Synonyms: TAK-659 HCl, 6-(((1R,2S)-2-aminocyclohexyl)amino)-7-fluoro-4-(1-methyl-1H-pyrazol-4-yl)-1,2-dihydro-3H-pyrrolo[3,4-c]pyridin-3-one hydrochloride

A863648-10MG A863648-1MG A863648-25MG A863648-5MG
77281-754EA 61 USD
77281-754 77281-756 77281-758 77281-760
TAK-659 Hydrochloride 98+%
TAK-659 Hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₂ClFN₆O
MDL Number: MFCD31544322
CAS Number: 1952251-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR