3-(1H-1,2,4-Triazol-1-yl)aniline 97%

Supplier: Ambeed
Warning

Synonyms: 3-(1,2,4-Triazol-1-yl)aniline

A863421-250MG A863421-1G
77281-752EA 50.47 USD
77281-752 77749-673
3-(1H-1,2,4-Triazol-1-yl)aniline 97%
3-(1H-1,2,4-Triazol-1-yl)aniline

Precaución: for Research Use Only

Formula: C₈H₈N₄
MW: 160.1759 g/mol
MDL Number: MFCD08271899
CAS Number: 176032-78-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR