BAY-876 99%

Supplier: Ambeed
Warning

Synonyms: N4-[1-[(4-cyanophenyl)methyl]-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl]-7-fluoro-2,4-Quinolinedicarboxamide, N4-(1-(4-Cyanobenzyl)-5-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)-7-fluoroquinoline-2,4-dicarboxamide

A863389-10MG A863389-1MG A863389-2MG A863389-5MG
77268-656EA 125.66 USD
77268-656 77268-658 77268-660 77268-662
BAY-876 99%
BAY-876

Precaución: for Research Use Only

Formula: C₂₄H₁₆F₄N₆O₂
MW: 496.42 g/mol
MDL Number: MFCD30377201
CAS Number: 1799753-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR