5,10,15,20-Tetra(4-methylphenyl)-21H,23H-porphine manganese(III) chloride 98%

Supplier: Ambeed
Warning

A863321-100MG A863321-1G A863321-250MG
77256-376EA 19 USD
77256-376 77256-378 77256-380
5,10,15,20-Tetra(4-methylphenyl)-21H,23H-porphine manganese(III) chloride 98%
5,10,15,20-Tetra(4-methylphenyl)-21H,23H-porphine manganese(III) chloride

Precaución: for Research Use Only

Formula: C₄₈H₃₈Cl₃MnN₄
MW: 832.13 g/mol
MDL Number: MFCD03453179
CAS Number: 43145-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR