Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromo-4-(trifluoromethyl)thiazole-5-carboxylate

A863279-1G A863279-250MG
77268-652EA 57.68 USD
77268-652 77268-654
Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate 98%
Ethyl-2-bromo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₅BrF₃NO₂S
MW: 304.09 g/mol
MDL Number: MFCD08166749
CAS Number: 72850-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR