4-(Cyanomethyl)benzeneboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetonitrile, 4-(Cyanomethyl)phenylboronic acid pinacol ester

A862905-100G A862905-1G A862905-25G A862905-5G
77294-040EA 540 USD
77294-040 77294-042 77294-044 77294-046
4-(Cyanomethyl)benzeneboronic acid pinacol ester 98%
4-(Cyanomethyl)benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₂
MW: 243.11 g/mol
MDL Number: MFCD02179459
CAS Number: 138500-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR