2-Methyl-3-(methylthio)furan 95%

Supplier: Ambeed
Danger

A861863-1G A861863-25G A861863-5G
77281-744EA 6 USD
77281-744 77281-746 77281-748
2-Methyl-3-(methylthio)furan 95%
2-Methyl-3-(methylthio)furan

Precaución: for Research Use Only

Formula: C₆H₈OS
MW: 128.19 g/mol
MDL Number: MFCD03701635
CAS Number: 63012-97-5
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR