4-Amino-6-(tert-butyl)pyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-(tert-butyl)-1,3-diazine, 6-(tert-Butyl)pyrimidin-4-amine

A861300-1G A861300-250MG A861300-5G
77294-028EA 127.72 USD
77294-028 77294-030 77294-032
4-Amino-6-(tert-butyl)pyrimidine 97%
4-Amino-6-(tert-butyl)pyrimidine

Precaución: for Research Use Only

Formula: C₈H₁₃N₃
MW: 151.209 g/mol
MDL Number: MFCD00234463
CAS Number: 3435-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR