2,4-Dihydroxy-7-methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one 98%

Supplier: Ambeed
Warning

Synonyms: DIMBOA

A861298-10MG A861298-1MG A861298-25MG A861298-5MG A861298-50MG A861298-100MG
77268-644EA 89.61 USD
77268-644 77268-646 77268-648 77268-650 77618-422 77749-668
2,4-Dihydroxy-7-methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one 98%
2,4-Dihydroxy-7-methoxy-2H-benzo[b][1,4]oxazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₉H₉NO₅
MW: 211.17 g/mol
MDL Number: MFCD01744500
CAS Number: 15893-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR