4'-Chloroacetanilide 98%

Supplier: Ambeed
Warning

A860823-100G A860823-25G A860823-500G A860823-5G
77294-022EA 53 USD
77294-022 77294-024 77294-026 77569-596
4'-Chloroacetanilide 98%
4'-Chloroacetanilide

Precaución: For research use only.

Formula: C₈H₈ClNO
MDL Number: MFCD00000612
CAS Number: 539-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR