2-Chloro-5-isopropoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

A860727-100MG A860727-250MG
77268-640EA 104.03 USD
77268-640 77268-642
2-Chloro-5-isopropoxy-3-nitropyridine 97%
2-Chloro-5-isopropoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₈H₉ClN₂O₃
MW: 216.62 g/mol
MDL Number: MFCD29761895
CAS Number: 1935294-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR