(R)-3,3'-Diiodo-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Danger

A860524-100MG A860524-1G A860524-250MG
77256-364EA 19 USD
77256-364 77256-366 77747-624
(R)-3,3'-Diiodo-[1,1'-binaphthalene]-2,2'-diol ≥98%, ee 99%
(R)-3,3'-Diiodo-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₂₀H₁₂I₂O₂
MW: 538.1 g/mol
MDL Number: MFCD25372833
CAS Number: 287111-93-7
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR