4-((3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)thiomorpholine 97%

Supplier: Ambeed
Warning

A859524-1G A859524-250MG A859524-5G
77244-536EA 80.34 USD
77244-536 77244-538 77244-540
4-((3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)thiomorpholine 97%
4-((3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulfonyl)thiomorpholine

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₄S₂
MW: 369.31 g/mol
MDL Number: MFCD08706095
CAS Number: 864754-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR