3-Bromoisobutyric acid 98%

Supplier: Ambeed
Danger

Synonyms: Captopril EP Impurity D, 3-Bromo-2-methylpropanoic acid, 3-Bromo-2-methylpropionic Acid

A859066-25G A859066-1G A859066-5G
77281-742EA 90.64 USD
77281-742 77618-414 77749-666
3-Bromoisobutyric acid 98%
3-Bromoisobutyric acid

Precaución: for Research Use Only

Formula: C₄H₇BrO₂
MW: 167.00 g/mol
MDL Number: MFCD01310530
CAS Number: 56970-78-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR