5-Methyl-2-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-nitro-m-tolyl ether, 3-Methoxy-4-nitrotoluene, 2-Methoxy-4-methyl-1-nitrobenzene, 1-Methoxy-3-methyl-6-nitrobenzene

A858706-100G A858706-10G A858706-1G A858706-25G A858706-5G
77256-354EA 395.52 USD
77256-354 77256-356 77256-358 77256-360 77256-362
5-Methyl-2-nitroanisole 98%
5-Methyl-2-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.16 g/mol
MDL Number: MFCD00007367
CAS Number: 38512-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR