2,3-Difluoro-6-iodobenzoic acid 95%

Supplier: Ambeed
Warning

A857987-1G A857987-250MG
77256-350EA 75.19 USD
77256-350 77256-352
2,3-Difluoro-6-iodobenzoic acid 95%
2,3-Difluoro-6-iodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂IO₂
MW: 283.99 g/mol
MDL Number: MFCD07368093
CAS Number: 333780-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR