3-(Methylthio)thiophene 97%

Supplier: Ambeed
Warning

A857179-1G A857179-250MG
77281-736EA 30.01 USD
77281-736 77281-738
3-(Methylthio)thiophene 97%
3-(Methylthio)thiophene

Precaución: for Research Use Only

Formula: C₅H₆S₂
MDL Number: MFCD00068009
CAS Number: 20731-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR